siRNA Details
| Gene Symbol: HGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000187.3 | NP_000178.2 | 4, 5 | 748 | ||
| XM_005247412.1 | XP_005247469.1 | 4, 5 | 748 | |||
| XM_005247413.1 | XP_005247470.1 | 4, 5 | 748 | |||
| XM_005247414.4 | XP_005247471.1 | 4, 5 | 748 | |||
| XM_011512746.1 | XP_011511048.1 | 4, 5 | 748 | |||
| XM_017006277.1 | XP_016861766.1 | 4, 5 | 361 | |||
| GenBank mRNA | AF045167.1 | - | 4, 5 | 455 | ||
| AK290782.1 | - | 4, 5 | 454 | |||
| AK313563.1 | - | 4, 5 | 732 | |||
| BC020792.1 | - | 4, 5 | 454 | 4716316 | ||
| BC071757.1 | - | 4, 5 | 386 | 6714217 | ||
| U63008.1 | - | 4, 5 | 417 | |||
| Z75048.1 | - | 4, 5 | 417 | |||

HGD
Genomic Map