siRNA Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276387.1 | NP_001263316.1 | 15 | 2409 | ||
| NM_010095.6 | NP_034225.1 | 15 | 2404 | |||
| GenBank mRNA | AK053013.1 | - | 15 | 2405 | ||
| AK139262.1 | - | 15 | 2116 | |||
| BC049188.1 | - | 14 | 1703 | 6312260 | ||
| BC050922.1 | - | 14 | 1926 | 6409737 | ||
| U71189.1 | - | 14 | 1589 | |||
| U82441.1 | - | 14 | 1458 | |||
| U92703.1 | - | 14 | 1809 | |||

Ebf2
Genomic Map