siRNA Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276387.1 | NP_001263316.1 | 7, 8 | 1518 | ||
| NM_010095.6 | NP_034225.1 | 7, 8 | 1513 | |||
| GenBank mRNA | AK053013.1 | - | 7, 8 | 1514 | ||
| AK139262.1 | - | 7, 8 | 1225 | |||
| BC049188.1 | - | 6, 7 | 812 | 6312260 | ||
| BC050922.1 | - | 6, 7 | 1035 | 6409737 | ||
| U71189.1 | - | 6, 7 | 698 | |||
| U82441.1 | - | 6, 7 | 567 | |||
| U92703.1 | - | 6, 7 | 918 | |||

Ebf2
Genomic Map