siRNA Details
| Gene Symbol: Ebf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113414.1 | NP_001106885.1 | 8 | 1187 | ||
| NM_001113415.1 | NP_001106886.1 | 8 | 1187 | |||
| NM_010096.3 | NP_034226.1 | 8 | 1187 | |||
| GenBank mRNA | AK138602.1 | - | 3 | 376 | ||
| AK220542.1 | - | 8 | 1187 | |||
| BC067018.1 | - | 9 | 942 | 6833120 | ||
| CF727275.1 | - | 2 | 52 | |||
| U82442.1 | - | 7 | 484 | |||
| U92702.1 | - | 8 | 728 | |||
| U92704.1 | - | 8 | 728 | |||

Ebf3
Genomic Map