siRNA Details
| Gene Symbol: Ebf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113414.1 | NP_001106885.1 | 3 | 851 | ||
| NM_001113415.1 | NP_001106886.1 | 3 | 851 | |||
| NM_010096.3 | NP_034226.1 | 3 | 851 | |||
| GenBank mRNA | AK220542.1 | - | 3 | 851 | ||
| BC067018.1 | - | 4 | 606 | 6833120 | ||
| U82442.1 | - | 2 | 148 | |||
| U92702.1 | - | 3 | 392 | |||
| U92704.1 | - | 3 | 392 | |||

Ebf3
Genomic Map