siRNA Details
| Gene Symbol: Ecm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252653.1 | NP_001239582.1 | 9 | 1409 | ||
| NM_007899.2 | NP_031925.2 | 9 | 1412 | |||
| GenBank mRNA | AK153009.1 | - | 9 | 1411 | ||
| AK153471.1 | - | 9 | 1411 | |||
| AK159334.1 | - | 9 | 1408 | |||
| AK169939.1 | - | 9 | 1414 | |||
| BC138693.1 | - | 9 | 1398 | 8861715 | ||
| BC145381.1 | - | 9 | 1395 | 9053939 | ||
| L33416.1 | - | 9 | 1405 | |||

Ecm1
Genomic Map