siRNA Details
| Gene Symbol: Edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010100.3 | NP_034230.1 | 4 | 455 | ||
| GenBank mRNA | AF160502.1 | - | 4 | 455 | ||
| BC068315.1 | - | 4 | 413 | 30098537 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Edar| Gene Symbol: Edar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010100.3 | NP_034230.1 | 4 | 455 | ||
| GenBank mRNA | AF160502.1 | - | 4 | 455 | ||
| BC068315.1 | - | 4 | 413 | 30098537 | ||