siRNA Details
| Gene Symbol: Edn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010104.3 | NP_034234.1 | 5 | 1169 | ||
| GenBank mRNA | AB081657.1 | - | 5 | 555 | ||
| AK040778.1 | - | 5 | 827 | |||
| AK152647.1 | - | 5 | 820 | |||
| AK153437.1 | - | 5 | 824 | |||
| BC029547.1 | - | 5 | 963 | 5059522 | ||
| D43775.1 | - | 5 | 880 | |||
| U35233.1 | - | 5 | 574 | |||

Edn1
Genomic Map