siRNA Details
| Gene Symbol: Eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007906.2 | NP_031932.1 | 3 | 525 | ||
| GenBank mRNA | BC018235.1 | - | 3 | 525 | 4504385 | |
| L26479.1 | - | 3 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eef1a2| Gene Symbol: Eef1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007906.2 | NP_031932.1 | 3 | 525 | ||
| GenBank mRNA | BC018235.1 | - | 3 | 525 | 4504385 | |
| L26479.1 | - | 3 | 291 | |||