siRNA Details
| Gene Symbol: Efna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010109.3 | NP_034239.1 | 2 | 592 | ||
| NM_207654.2 | NP_997537.1 | 2 | 592 | |||
| GenBank mRNA | BC040218.1 | - | 2 | 592 | 4954352 | |
| U90664.1 | - | 2 | 419 | |||
| U90665.1 | - | 2 | 419 | |||

Efna5
Genomic Map