siRNA Details
Gene Symbol: Efnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010111.5 | NP_034241.2 | 2 | 310 | ||
GenBank mRNA | AK132512.1 | - | 2 | 777 | ||
AK132845.1 | - | 2 | 603 | |||
AK134108.1 | - | 2 | 777 | |||
AK142203.1 | - | 2 | 780 | |||
AK142283.1 | - | 2 | 780 | |||
AK143059.1 | - | 2 | 777 | |||
BC057009.1 | - | 2 | 598 | 6827408 | ||
CT010381.1 | - | 2 | 151 | |||
L38847.1 | - | 2 | 311 | |||
U16819.1 | - | 2 | 179 | |||
U30244.1 | - | 2 | 148 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
