siRNA Details
| Gene Symbol: Klk1b22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010114.1 | NP_034244.1 | 4 | 580 | ||
| GenBank mRNA | BC106155.1 | - | 4 | 584 | 4189167 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klk1b22| Gene Symbol: Klk1b22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010114.1 | NP_034244.1 | 4 | 580 | ||
| GenBank mRNA | BC106155.1 | - | 4 | 584 | 4189167 | |