siRNA Details
Gene Symbol: Klk1b22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010114.1 | NP_034244.1 | 3, 4 | 503 | ||
GenBank mRNA | BC106155.1 | - | 3, 4 | 507 | 4189167 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klk1b22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010114.1 | NP_034244.1 | 3, 4 | 503 | ||
GenBank mRNA | BC106155.1 | - | 3, 4 | 507 | 4189167 |