siRNA Details
| Gene Symbol: Egr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010118.3 | NP_034248.2 | 2 | 1503 | ||
| GenBank mRNA | AK153882.1 | - | 2 | 1503 | ||
| AK155585.1 | - | 2 | 1504 | |||
| BC009093.1 | - | 2 | 1268 | 3157863 | ||
| X06746.1 | - | 3 | 1393 | |||

Egr2
Genomic Map