siRNA Details
| Gene Symbol: Ehf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007914.3 | NP_031940.1 | 3 | 292 | ||
| GenBank mRNA | AF035527.1 | - | 2 | 290 | ||
| AK028651.1 | - | 3 | 263 | |||
| BC005520.1 | - | 3 | 254 | 3494644 | ||
| BC006789.1 | - | 3 | 274 | 3589528 | ||

Ehf
Genomic Map