siRNA Details
| Gene Symbol: Eif3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010123.3 | NP_034253.3 | 2 | 392 | ||
| GenBank mRNA | AK082006.1 | - | 2 | 358 | ||
| AK145132.1 | - | 2 | 400 | |||
| AK145629.1 | - | 2 | 357 | |||
| AK149482.1 | - | 2 | 345 | |||
| AK161634.1 | - | 2 | 349 | |||
| AK167010.1 | - | 2 | 359 | |||
| AK172899.1 | - | 2 | 329 | |||
| U14172.1 | - | 2 | 342 | |||

Eif3a
Genomic Map