siRNA Details
| Gene Symbol: Eif4ebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010124.2 | NP_034254.1 | 1, 2 | 273 | ||
| GenBank mRNA | AK133912.1 | - | 1, 2 | 161 | ||
| AK148374.1 | - | 312 | ||||
| AK210066.1 | - | 11 | ||||
| AK216468.1 | - | 11 | ||||
| AK217241.1 | - | 11 | ||||
| AK217805.1 | - | 11 | ||||
| BC015082.1 | - | 2, 3 | 254 | 4218820 | ||
| BC119539.2 | - | 2, 3 | 237 | 40056467 | ||
| U75530.1 | - | 1, 2 | 270 | |||

Eif4ebp2
Genomic Map