siRNA Details
| Gene Symbol: Elk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007923.2 | NP_031949.2 | 2 | 500 | ||
| GenBank mRNA | AK054075.1 | - | 2 | 792 | ||
| AK156537.1 | - | 2 | 498 | |||
| BC144844.1 | - | 2 | 250 | 9053392 | ||
| BC145795.1 | - | 2 | 250 | 40131098 | ||
| FN434116.1 | - | 1 | 97 | |||
| FN434117.1 | - | 1 | 97 | |||
| FN434118.1 | - | 1 | 97 | |||
| Z36885.1 | - | 3 | 445 | |||

Elk4
Genomic Map