siRNA Details
| Gene Symbol: Emp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288627.1 | NP_001275556.1 | 5 | 655 | ||
| NM_001288628.1 | NP_001275557.1 | 6 | 724 | |||
| NM_010128.4 | NP_034258.1 | 5 | 613 | |||
| GenBank mRNA | AK135453.1 | - | 3 | 328 | ||
| AK157366.1 | - | 5 | 531 | |||
| AK159752.1 | - | 5 | 531 | |||
| AK168806.1 | - | 5 | 505 | |||
| AK171061.1 | - | 5 | 531 | |||
| AK171675.1 | - | 5 | 531 | |||
| AK172607.1 | - | 5 | 531 | |||
| AK172705.1 | - | 5 | 531 | |||
| BC034257.1 | - | 6 | 624 | 3157463 | ||
| BC046299.1 | - | 5 | 613 | 6486958 | ||
| BC050899.1 | - | 5 | 655 | 6402273 | ||
| BC138491.1 | - | 5 | 474 | 8861511 | ||
| CT010209.1 | - | 4 | 408 | |||
| U25633.1 | - | 5 | 497 | |||
| X98403.1 | - | 5 | 590 | |||

Emp1
Genomic Map