siRNA Details
| Gene Symbol: Emp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007929.2 | NP_031955.2 | 3 | 390 | ||
| GenBank mRNA | AF083876.1 | - | 3 | 257 | ||
| AF346627.1 | - | 3 | 332 | |||
| AK076194.1 | - | 3 | 369 | |||
| AK077260.1 | - | 3 | 392 | |||
| BC031646.1 | - | 2 | 183 | |||
| BC041670.1 | - | 3 | 293 | 3663158 | ||

Emp2
Genomic Map