siRNA Details
| Gene Symbol: Adgre1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010130.4 | NP_034260.1 | 7, 8 | 852 | ||
| GenBank mRNA | AK133202.1 | - | 7, 8 | 828 | ||
| AK149898.1 | - | 7, 8 | 838 | |||
| AK151303.1 | - | 7, 8 | 824 | |||
| AK151682.1 | - | 7, 8 | 844 | |||
| AK155862.1 | - | 7, 8 | 803 | |||
| BC075688.1 | - | 7, 8 | 824 | 30609706 | ||
| U66888.1 | - | 7, 8 | 839 | |||
| U66889.1 | - | 7, 8 | 839 | |||
| U66891.1 | - | 5, 6 | 563 | |||
| U66892.1 | - | 4, 5 | 416 | |||
| X93328.1 | - | 7, 8 | 848 | |||
More Information
Set Membership: |
|

Adgre1
Genomic Map