siRNA Details
| Gene Symbol: Emx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010132.2 | NP_034262.2 | 3 | 1150 | ||
| GenBank mRNA | AK132093.1 | - | 3 | 938 | ||
| AY117415.1 | - | 3 | 1468 | |||
| X68882.1 | - | 2 | 162 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Emx2| Gene Symbol: Emx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010132.2 | NP_034262.2 | 3 | 1150 | ||
| GenBank mRNA | AK132093.1 | - | 3 | 938 | ||
| AY117415.1 | - | 3 | 1468 | |||
| X68882.1 | - | 2 | 162 | |||