siRNA Details
| Gene Symbol: Enc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007930.4 | NP_031956.3 | 2 | 496 | ||
| GenBank mRNA | AK008709.1 | - | 2 | 312 | ||
| AK044906.1 | - | 2 | 496 | |||
| AK049187.1 | - | 2 | 491 | |||
| AK132173.1 | - | 2 | 496 | |||
| BC049186.1 | - | 2 | 127 | 6485551 | ||
| BC058098.1 | - | 2 | 431 | 6824970 | ||
| U65079.1 | - | 2 | 348 | |||

Enc1
Genomic Map