siRNA Details
| Gene Symbol: Eng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146348.1 | NP_001139820.1 | 3 | 662 | ||
| NM_001146350.1 | NP_001139822.1 | 3 | 665 | |||
| NM_007932.2 | NP_031958.2 | 3 | 665 | |||
| GenBank mRNA | AK151302.1 | - | 3 | 635 | ||
| AK196985.1 | - | 1 | 84 | |||
| AY679531.1 | - | 3 | 665 | |||
| BC029080.1 | - | 3 | 579 | 4168250 | ||
| S69407.1 | - | 4 | 530 | |||
| X77952.1 | - | 3 | 665 | |||

Eng
Genomic Map