siRNA Details
| Gene Symbol: Epas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010137.3 | NP_034267.3 | 13 | 2535 | ||
| GenBank mRNA | AF045160.1 | - | 13 | 2299 | ||
| AK136911.1 | - | 13 | 2539 | |||
| AK141185.1 | - | 13 | 2270 | |||
| AK158430.1 | - | 13 | 2699 | |||
| BC057870.1 | - | 13 | 2257 | 5032291 | ||
| D89787.1 | - | 13 | 2497 | |||
| U81983.1 | - | 13 | 2218 | |||

Epas1
Genomic Map