siRNA Details
| Gene Symbol: Epha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023580.4 | NP_076069.2 | 15, 16 | 2562 | ||
| GenBank mRNA | AF131197.1 | - | 15, 16 | 2504 | ||
| AK028478.1 | - | 15, 16 | 2563 | |||
| BC071215.1 | - | 15, 16 | 2526 | 4196138 | ||

Epha1
Genomic Map