siRNA Details
| Gene Symbol: Epha4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007936.3 | NP_031962.2 | 8, 9 | 1774 | ||
| GenBank mRNA | AK028509.1 | - | 8, 9 | 1767 | ||
| AK147698.1 | - | 8, 9 | 1770 | |||
| BC004782.1 | - | 8, 9 | 1689 | 3588285 | ||
| BC052164.1 | - | 8, 9 | 1746 | 6512978 | ||
| S57168.1 | - | 8, 9 | 1774 | |||
| X65138.1 | - | 8, 9 | 1774 | |||

Epha4
Genomic Map