siRNA Details
| Gene Symbol: Ephb6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146351.1 | NP_001139823.1 | 18 | 3499 | ||
| NM_007680.4 | NP_031706.3 | 18 | 3495 | |||
| GenBank mRNA | AK076442.1 | - | 18 | 3322 | ||
| AK087423.1 | - | 18 | 3410 | |||
| AK163823.1 | - | 9 | 1217 | |||
| BB833527.1 | - | 2 | 199 | |||
| BC031924.1 | - | 17 | 3107 | 4911959 | ||
| L77867.1 | - | 18 | 3189 | |||

Ephb6
Genomic Map