siRNA Details
| Gene Symbol: Epn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252188.1 | NP_001239117.1 | 10 | 1940 | ||
| NM_001252189.1 | NP_001239118.1 | 8 | 1696 | |||
| NM_010148.3 | NP_034278.1 | 9 | 1732 | |||
| GenBank mRNA | AF057286.1 | - | 7 | 1187 | ||
| AK036331.1 | - | 8 | 1670 | |||
| AK139234.1 | - | 10 | 1928 | |||
| AK139793.1 | - | 10 | 1863 | |||
| AK160782.1 | - | 8 | 1697 | |||
| AK173093.1 | - | 10 | 1916 | |||
| BC039138.1 | - | 9 | 1696 | 2647379 | ||

Epn2
Genomic Map