siRNA Details
| Gene Symbol: Nr2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010151.2 | NP_034281.2 | 3 | 1568 | ||
| GenBank mRNA | AK148880.1 | - | 3 | 1905 | ||
| BC048695.1 | - | 3 | 1254 | |||
| BC108408.1 | - | 3 | 1457 | 30245025 | ||
| U07625.1 | - | 3 | 1170 | |||
| X74134.1 | - | 3 | 1468 | |||

Nr2f1
Genomic Map