siRNA Details
| Gene Symbol: Nr2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010151.2 | NP_034281.2 | 2 | 1121 | ||
| GenBank mRNA | AK014008.1 | - | 1 | 208 | ||
| AK148880.1 | - | 2 | 1458 | |||
| BC048695.1 | - | 2 | 807 | |||
| BC108408.1 | - | 2 | 1010 | 30245025 | ||
| U07625.1 | - | 2 | 723 | |||
| X74134.1 | - | 2 | 1021 | |||

Nr2f1
Genomic Map