siRNA Details
| Gene Symbol: Erbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010153.1 | NP_034283.1 | 19 | 2221 | ||
| GenBank mRNA | AY686636.1 | - | 19 | 2221 | ||
| BC049279.1 | - | 33 | ||||
| BC106091.1 | - | 2372 | 30844039 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erbb3| Gene Symbol: Erbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010153.1 | NP_034283.1 | 19 | 2221 | ||
| GenBank mRNA | AY686636.1 | - | 19 | 2221 | ||
| BC049279.1 | - | 33 | ||||
| BC106091.1 | - | 2372 | 30844039 | |||