siRNA Details
| Gene Symbol: Erbb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010153.1 | NP_034283.1 | 22, 23 | 2687 | ||
| GenBank mRNA | AY686636.1 | - | 22, 23 | 2687 | ||
| BC029028.1 | - | 202 | ||||
| BC036180.1 | - | 291 | ||||
| BC049279.1 | - | 499 | ||||
| BC106091.1 | - | 2838 | 30844039 | |||

Erbb3
Genomic Map