siRNA Details
| Gene Symbol: Amz2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252193.1 | NP_001239122.1 | 8 | 1462 | ||
| NM_025275.4 | NP_079551.3 | 9 | 1281 | |||
| GenBank mRNA | AJ879914.1 | - | 6 | 978 | ||
| AK004062.1 | - | 9 | 1241 | |||
| AK075748.1 | - | 9 | 1221 | |||
| BC025087.1 | - | 8 | 1456 | 2649126 | ||

Amz2
Genomic Map