siRNA Details
Gene Symbol: Esr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010157.3 | NP_034287.3 | 9 | 1950 | ||
NM_207707.1 | NP_997590.1 | 10 | 2004 | |||
GenBank mRNA | AK054290.1 | - | 10 | 2002 | ||
AK054301.1 | - | 8 | 1812 | |||
AK054413.1 | - | 10 | 2005 | |||
AY054413.1 | - | 9 | 1465 | |||
BC141075.1 | - | 8 | 1652 | 9055687 | ||
BC145329.1 | - | 8 | 1652 | 9053885 | ||
U81451.1 | - | 8 | 1422 |
