siRNA Details
| Gene Symbol: Esr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010157.3 | NP_034287.3 | 2 | 757 | ||
| NM_207707.1 | NP_997590.1 | 2 | 757 | |||
| GenBank mRNA | AK054290.1 | - | 2 | 755 | ||
| AK054301.1 | - | 2 | 758 | |||
| AK054413.1 | - | 2 | 758 | |||
| AY054413.1 | - | 1 | 218 | |||
| BC141075.1 | - | 1 | 459 | 9055687 | ||
| BC145329.1 | - | 1 | 459 | 9053885 | ||
| U81451.1 | - | 1 | 229 | |||

Esr2
Genomic Map