siRNA Details
| Gene Symbol: Etv6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK035231.1 | - | 5 | 790 | ||
| AK078773.1 | - | 6 | 983 | |||
| AK154569.1 | - | 6 | 1071 | |||
| AK155150.1 | - | 6 | 1256 | |||
| AK155752.1 | - | 5 | 806 | |||
| AK157265.1 | - | 6 | 925 | |||
| BC052163.1 | - | 6 | 944 | 6513848 | ||

Etv6
Genomic Map