siRNA Details
| Gene Symbol: Evi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007964.2 | NP_031990.2 | 4 | 688 | ||
| GenBank mRNA | AK134294.1 | - | 4 | 889 | ||
| AK164899.1 | - | 1 | 13 | |||
| AK165332.1 | - | 5 | 681 | |||
| AK165885.1 | - | 4 | 641 | |||
| AK192278.1 | - | 2 | 178 | |||
| AK216641.1 | - | 2 | 364 | |||
| BC030056.1 | - | 5 | 695 | |||
| BI737814.1 | - | 4 | 612 | |||
| U53586.1 | - | 4 | 688 | |||

Evi5
Genomic Map