siRNA Details
| Gene Symbol: Evi5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007964.2 | NP_031990.2 | 8 | 1123 | ||
| GenBank mRNA | AK134294.1 | - | 8 | 1324 | ||
| AK164899.1 | - | 5 | 448 | |||
| AK165332.1 | - | 10 | 1150 | |||
| AK165885.1 | - | 8 | 1076 | |||
| AK218636.1 | - | 2 | 109 | |||
| BC030056.1 | - | 9 | 1130 | |||
| U53586.1 | - | 8 | 1123 | |||

Evi5
Genomic Map