siRNA Details
| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007969.4 | NP_031995.3 | 2 | 246 | ||
| GenBank mRNA | BE310436.1 | - | 2 | 107 | ||
| EH105860.1 | - | 2 | 273 | |||
| EH109450.1 | - | 2 | 152 | |||
| X93037.1 | - | 2 | 144 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc18| Gene Symbol: Wfdc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007969.4 | NP_031995.3 | 2 | 246 | ||
| GenBank mRNA | BE310436.1 | - | 2 | 107 | ||
| EH105860.1 | - | 2 | 273 | |||
| EH109450.1 | - | 2 | 152 | |||
| X93037.1 | - | 2 | 144 | |||