siRNA Details
| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010164.2 | NP_034294.2 | 5, 6 | 874 | ||
| GenBank mRNA | AK153788.1 | - | 5, 6 | 879 | ||
| BC060260.1 | - | 5, 6 | 874 | 6848408 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eya1| Gene Symbol: Eya1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010164.2 | NP_034294.2 | 5, 6 | 874 | ||
| GenBank mRNA | AK153788.1 | - | 5, 6 | 879 | ||
| BC060260.1 | - | 5, 6 | 874 | 6848408 | ||