siRNA Details
| Gene Symbol: Eya3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010166.3 | NP_034296.2 | 13, 14 | 1222 | ||
| NM_210071.2 | NP_997592.1 | 12, 13 | 1183 | |||
| NM_211357.2 | NP_997596.1 | 10, 11 | 1002 | |||
| GenBank mRNA | AK144653.1 | - | 12, 13 | 1174 | ||
| BC052860.1 | - | 11, 12 | 986 | |||
| BC063259.1 | - | 12, 13 | 1111 | 6406090 | ||
| U61112.1 | - | 9, 10 | 887 | |||
| U81604.1 | - | 13, 14 | 1190 | |||

Eya3
Genomic Map