siRNA Details
| Gene Symbol: Eya3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010166.3 | NP_034296.2 | 8, 9 | 664 | ||
| NM_210071.2 | NP_997592.1 | 7, 8 | 625 | |||
| NM_211357.2 | NP_997596.1 | 5, 6 | 444 | |||
| GenBank mRNA | AK144653.1 | - | 7, 8 | 616 | ||
| AK164591.1 | - | 7, 8 | 625 | |||
| BC052860.1 | - | 6, 7 | 428 | |||
| BC063259.1 | - | 7, 8 | 553 | 6406090 | ||
| BY730487.1 | - | 7, 8 | 625 | |||
| U61112.1 | - | 4, 5 | 329 | |||
| U81604.1 | - | 8, 9 | 632 | |||

Eya3
Genomic Map