siRNA Details
| Gene Symbol: Ezh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB004817.1 | - | 7 | 628 | ||
| AF483490.1 | - | 5 | 517 | |||
| AF483491.1 | - | 5 | 517 | |||
| AK045374.1 | - | 7 | 616 | |||
| AK129004.1 | - | 6 | 588 | |||
| AK138942.1 | - | 7 | 611 | |||
| AK140694.1 | - | 7 | 635 | |||
| AK154565.1 | - | 7 | 608 | |||
| AK164192.1 | - | 7 | 627 | |||
| BC007135.1 | - | 6 | 588 | 3600884 | ||
| U60453.1 | - | 7 | 616 | |||

Ezh1
Genomic Map