siRNA Details
| Gene Symbol: F10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242368.1 | NP_001229297.1 | 7, 8 | 1199 | ||
| NM_007972.4 | NP_031998.3 | 7, 8 | 1009 | |||
| GenBank mRNA | AF087644.1 | - | 6, 7 | 746 | ||
| AJ222677.1 | - | 6, 7 | 772 | |||
| AK149449.1 | - | 6, 7 | 793 | |||
| AK154100.1 | - | 7, 8 | 1009 | |||
| AK154572.1 | - | 7, 8 | 1126 | |||
| AK170282.1 | - | 7, 8 | 1102 | |||
| AK171096.1 | - | 7, 8 | 1009 | |||
| BC050219.1 | - | 7, 8 | 1185 | 30018889 | ||

F10
Genomic Map