siRNA Details
| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010170.4 | NP_034300.3 | 2 | 658 | ||
| GenBank mRNA | AK089444.1 | - | 2 | 622 | ||
| AK135212.1 | - | 2 | 725 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F2rl2| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010170.4 | NP_034300.3 | 2 | 658 | ||
| GenBank mRNA | AK089444.1 | - | 2 | 622 | ||
| AK135212.1 | - | 2 | 725 | |||