siRNA Details
| Gene Symbol: F2rl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010170.4 | NP_034300.3 | 2 | 337 | ||
| GenBank mRNA | AK089444.1 | - | 2 | 301 | ||
| AK135212.1 | - | 2 | 404 | |||
| BC140985.1 | - | 2 | 292 | 9055597 | ||
| U92972.1 | - | 2 | 345 | |||

F2rl2
Genomic Map