siRNA Details
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137521.1 | - | 13 | 2861 | ||
AK137537.1 | - | 13 | 2838 | |||
AK139240.1 | - | 13 | 2855 | |||
U52925.1 | - | 13 | 2736 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137521.1 | - | 13 | 2861 | ||
AK137537.1 | - | 13 | 2838 | |||
AK139240.1 | - | 13 | 2855 | |||
U52925.1 | - | 13 | 2736 |