siRNA Details
| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK149372.1 | - | 8 | 1199 | ||
| BC125617.1 | - | 8 | 1197 | |||
| BC132393.1 | - | 8 | 1197 | |||
| M26236.1 | - | 3 | 661 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

F9| Gene Symbol: F9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK149372.1 | - | 8 | 1199 | ||
| BC125617.1 | - | 8 | 1197 | |||
| BC132393.1 | - | 8 | 1197 | |||
| M26236.1 | - | 3 | 661 | |||