siRNA Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004897.1 | - | 6 | 589 | ||
| AK145900.1 | - | 6 | 716 | |||
| AK149406.1 | - | 6 | 594 | |||
| AK152772.1 | - | 6 | 594 | |||
| AK153050.1 | - | 6 | 722 | |||
| AK158388.1 | - | 5 | 2502 | |||
| AK161189.1 | - | 6 | 568 | |||
| AK168078.1 | - | 6 | 594 | |||
| BC056644.1 | - | 6 | 578 | 5098645 | ||
| U15977.1 | - | 5 | 507 | |||

Acsl1
Genomic Map